siRNA Details
| Gene Symbol: Gale | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080783.2 | NP_542961.2 | 7, 8 | 747 | ||
| XM_006239134.2 | XP_006239196.1 | 7, 8 | 1712 | |||
| XM_008764197.1 | XP_008762419.1 | 7, 8 | 978 | |||
| GenBank mRNA | BC097293.1 | - | 7, 8 | 747 | 7443707 | |
| X53949.1 | - | 6, 7 | 725 | |||

Gale
Genomic Map