siRNA Details
| Gene Symbol: Slc9a3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053811.3 | NP_446263.2 | 6, 7 | 932 | ||
| XM_006245893.2 | XP_006245955.1 | 5, 6 | 1305 | |||
| GenBank mRNA | AF259898.1 | - | 6, 7 | 791 | ||
| AF294257.1 | - | 6, 7 | 1019 | |||

Slc9a3r2
Genomic Map