siRNA Details
Gene Symbol: Cplx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053878.1 | NP_446330.1 | 3 | 486 | ||
XM_006253596.2 | XP_006253658.1 | 3 | 522 | |||
XM_006253597.2 | XP_006253659.1 | 4 | 765 | |||
XM_008771482.1 | XP_008769704.1 | 4 | 765 | |||
XM_008771483.1 | XP_008769705.1 | 4 | 683 | |||
GenBank mRNA | D70816.1 | - | 3 | 486 | ||
U35099.1 | - | 4 | 474 |
