siRNA Details
| Gene Symbol: Napa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080585.1 | NP_542152.1 | 4, 5 | 380 | ||
| GenBank mRNA | BC063156.1 | - | 4, 5 | 439 | 6921187 | |
| FQ213217.1 | - | 4, 5 | 384 | |||
| FQ216375.1 | - | 4, 5 | 698 | |||
| X89968.1 | - | 4, 5 | 380 | |||

Napa
Genomic Map