siRNA Details
| Gene Symbol: Cacng5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080693.1 | NP_542424.1 | 2 | 269 | ||
| GenBank mRNA | AF361342.1 | - | 2 | 269 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacng5| Gene Symbol: Cacng5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080693.1 | NP_542424.1 | 2 | 269 | ||
| GenBank mRNA | AF361342.1 | - | 2 | 269 | ||