siRNA Details
| Gene Symbol: Dynll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080697.2 | NP_542428.1 | 2 | 271 | ||
| XM_006247077.2 | XP_006247139.1 | 2 | 50 | |||
| XM_008767940.1 | XP_008766162.1 | 2 | 41 | |||
| GenBank mRNA | AY034383.1 | - | 2 | 165 | ||
| BC061874.1 | - | 2 | 271 | 5598327 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001168471, NM_026556, NM_001168472, BC011289, BY310878, and others.

Dynll2
Genomic Map