siRNA Details
| Gene Symbol: Anxa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130416.2 | NP_569100.2 | 12, 13 | 1441 | ||
| XM_006251624.2 | XP_006251686.1 | 13, 14 | 1468 | |||
| XM_006251625.2 | XP_006251687.1 | 13, 14 | 3267 | |||
| XM_006251626.2 | XP_006251688.1 | 14, 15 | 3348 | |||
| XM_006251627.2 | XP_006251689.1 | 12, 13 | 1400 | |||
| XM_008770484.1 | XP_008768706.1 | 13, 14 | 1649 | |||
| XM_008770485.1 | XP_008768707.1 | 12, 13 | 1582 | |||
| GenBank mRNA | AF280423.1 | - | 1433 | |||
| BC070896.1 | - | 12, 13 | 1441 | 7099644 | ||

Anxa7
Genomic Map