siRNA Details
| Gene Symbol: Runx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130425.1 | NP_569109.1 | 3 | 506 | ||
| GenBank mRNA | AF421886.1 | - | 3 | 506 | ||
| FQ230848.1 | - | 255 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Runx3| Gene Symbol: Runx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130425.1 | NP_569109.1 | 3 | 506 | ||
| GenBank mRNA | AF421886.1 | - | 3 | 506 | ||
| FQ230848.1 | - | 255 | ||||