siRNA Details
| Gene Symbol: Hspb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130431.1 | NP_569115.1 | 2 | 413 | ||
| XM_006243027.2 | XP_006243089.1 | 2 | 528 | |||
| Gene Symbol: Cryab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ220032.1 | - | 154 | |||

Hspb2
Genomic Map