siRNA Details
| Gene Symbol: Phkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255269.2 | XP_006255331.1 | 30, 31 | 3205 | ||
| XM_006255270.2 | XP_006255332.1 | 30, 31 | 3205 | |||
| XM_008772410.1 | XP_008770632.1 | 30, 31 | 3917 | |||
| GenBank mRNA | FQ234320.1 | - | 5, 6 | 853 | ||

Phkb
Genomic Map