siRNA Details
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133393.2 | NP_596884.1 | 3 | 549 | ||
GenBank mRNA | AB054539.1 | - | 3 | 490 | ||
BC070933.1 | - | 3 | 549 | 7097855 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133393.2 | NP_596884.1 | 3 | 549 | ||
GenBank mRNA | AB054539.1 | - | 3 | 490 | ||
BC070933.1 | - | 3 | 549 | 7097855 |