siRNA Details
| Gene Symbol: Lfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133393.2 | NP_596884.1 | 3 | 610 | ||
| GenBank mRNA | AB054539.1 | - | 3 | 551 | ||
| BC070933.1 | - | 3 | 610 | 7097855 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lfng| Gene Symbol: Lfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133393.2 | NP_596884.1 | 3 | 610 | ||
| GenBank mRNA | AB054539.1 | - | 3 | 551 | ||
| BC070933.1 | - | 3 | 610 | 7097855 | ||