siRNA Details
Gene Symbol: Mpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133561.1 | NP_598245.1 | 3, 4 | 246 | ||
GenBank mRNA | AF304429.1 | - | 3, 4 | 246 | ||
BC097287.1 | - | 3, 4 | 262 | 7445460 | ||
FQ219877.1 | - | 3, 4 | 300 |
