siRNA Details
Gene Symbol: Tgoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138840.2 | NP_620195.1 | 2 | 960 | ||
GenBank mRNA | BC098512.1 | - | 2 | 960 | 7382894 | |
X53565.1 | - | 2 | 958 | |||
X64600.1 | - | 2 | 958 |
