siRNA Details
| Gene Symbol: Gpr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138841.1 | NP_620196.1 | 3 | 825 | ||
| GenBank mRNA | AF208288.1 | - | 3 | 825 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr26| Gene Symbol: Gpr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138841.1 | NP_620196.1 | 3 | 825 | ||
| GenBank mRNA | AF208288.1 | - | 3 | 825 | ||
Set Membership: |
|