siRNA Details
Gene Symbol: Gpr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138841.1 | NP_620196.1 | 1 | 586 | ||
GenBank mRNA | AF208288.1 | - | 1 | 586 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
