siRNA Details
| Gene Symbol: Acot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138907.2 | NP_620262.2 | 1 | 585 | ||
| XM_006240320.2 | XP_006240382.1 | 1 | 669 | |||
| GenBank mRNA | AB010429.1 | - | 1 | 595 | ||
| BC072540.1 | - | 1 | 585 | 7104319 | ||
| Y09333.1 | - | 1 | 526 | |||

Acot2
Genomic Map