siRNA Details
Gene Symbol: Nos1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138922.1 | NP_620277.1 | 10 | 1749 | ||
GenBank mRNA | AF037071.1 | - | 10 | 1749 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nos1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138922.1 | NP_620277.1 | 10 | 1749 | ||
GenBank mRNA | AF037071.1 | - | 10 | 1749 |