siRNA Details
| Gene Symbol: Adcyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016989.2 | NP_058685.1 | 3, 4 | 544 | ||
| XM_006245672.2 | XP_006245734.1 | 4, 5 | 1240 | |||
| XM_006245673.2 | XP_006245735.1 | 4, 5 | 1616 | |||
| XM_006245674.2 | XP_006245736.1 | 4, 5 | 1569 | |||
| XM_008767418.1 | XP_008765640.1 | 4, 5 | 1193 | |||
| XM_008767419.1 | XP_008765641.1 | 2, 3 | 590 | |||

Adcyap1
Genomic Map