siRNA Details
| Gene Symbol: Bcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016993.1 | NP_058689.1 | 1 | 251 | ||
| GenBank mRNA | AF512835.1 | - | 1 | 17 | ||
| L14680.1 | - | 1 | 251 | |||
| U34964.1 | - | 1 | 17 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl2| Gene Symbol: Bcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016993.1 | NP_058689.1 | 1 | 251 | ||
| GenBank mRNA | AF512835.1 | - | 1 | 17 | ||
| L14680.1 | - | 1 | 251 | |||
| U34964.1 | - | 1 | 17 | |||