siRNA Details
| Gene Symbol: Hagh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033349.2 | NP_203500.2 | 9 | 934 | ||
| XM_006245899.1 | XP_006245961.1 | 9 | 911 | |||
| XM_006245900.1 | XP_006245962.1 | 8 | 825 | |||
| GenBank mRNA | BC097301.1 | - | 9 | 886 | ||

Hagh
Genomic Map