siRNA Details
| Gene Symbol: Ina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019128.4 | NP_062001.1 | 3 | 2570 | ||
| GenBank mRNA | X52017.1 | - | 3 | 1297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ina| Gene Symbol: Ina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019128.4 | NP_062001.1 | 3 | 2570 | ||
| GenBank mRNA | X52017.1 | - | 3 | 1297 | ||