siRNA Details
| Gene Symbol: Bmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139258.1 | NP_640351.1 | 3 | 332 | ||
| XM_006234723.2 | XP_006234785.1 | 3 | 370 | |||
| XM_006234724.2 | XP_006234786.1 | 3 | 373 | |||
| XM_008762079.1 | XP_008760301.1 | 3 | 371 | |||
| GenBank mRNA | AF506761.1 | - | 3 | 332 | ||

Bmf
Genomic Map