siRNA Details
| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139259.2 | NP_640352.1 | 2 | 203 | ||
| XM_006243921.2 | XP_006243983.1 | 2 | 354 | |||
| GenBank mRNA | AF497263.1 | - | 2 | 207 | ||
| AF497264.1 | - | 2 | 202 | |||
| AF534395.1 | - | 1 | 19 | |||

Nradd
Genomic Map