siRNA Details
| Gene Symbol: Il3ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248989.2 | XP_006249051.1 | 8, 9 | 3305 | ||
| GenBank mRNA | AF030243.1 | - | 3, 4 | 509 | ||
| BP475867.1 | - | 4, 5 | 391 | |||
| FQ214452.1 | - | 5, 6 | 883 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il3ra| Gene Symbol: Il3ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248989.2 | XP_006249051.1 | 8, 9 | 3305 | ||
| GenBank mRNA | AF030243.1 | - | 3, 4 | 509 | ||
| BP475867.1 | - | 4, 5 | 391 | |||
| FQ214452.1 | - | 5, 6 | 883 | |||