siRNA Details
| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145094.2 | NP_659562.2 | 4 | 303 | ||
| GenBank mRNA | AF254800.1 | - | 4 | 252 | ||
| BC072698.1 | - | 4 | 303 | 7105732 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab31| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145094.2 | NP_659562.2 | 4 | 303 | ||
| GenBank mRNA | AF254800.1 | - | 4 | 252 | ||
| BC072698.1 | - | 4 | 303 | 7105732 | ||