siRNA Details
| Gene Symbol: Prl4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017036.2 | NP_058732.2 | 5 | 597 | ||
| GenBank mRNA | BC097332.1 | - | 5 | 597 | 7443660 | |
| FQ228460.1 | - | 5 | 632 | |||
| M13750.1 | - | 5 | 580 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl4a1| Gene Symbol: Prl4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017036.2 | NP_058732.2 | 5 | 597 | ||
| GenBank mRNA | BC097332.1 | - | 5 | 597 | 7443660 | |
| FQ228460.1 | - | 5 | 632 | |||
| M13750.1 | - | 5 | 580 | |||