siRNA Details
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145681.1 | NP_663714.1 | 1, 2 | 160 | ||
GenBank mRNA | AY115578.1 | - | 1, 2 | 160 | ||
EF030546.1 | - | 1, 2 | 151 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145681.1 | NP_663714.1 | 1, 2 | 160 | ||
GenBank mRNA | AY115578.1 | - | 1, 2 | 160 | ||
EF030546.1 | - | 1, 2 | 151 |