siRNA Details
| Gene Symbol: Prph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012633.2 | NP_036765.2 | 9 | 1431 | ||
| XM_006257321.2 | XP_006257383.1 | 8 | 2313 | |||
| XM_006257322.2 | XP_006257384.1 | 9 | 2215 | |||
| XM_008765669.1 | XP_008763891.1 | 8 | 2312 | |||
| GenBank mRNA | AF031878.1 | - | 9 | 1427 | ||
| BC100656.1 | - | 9 | 1431 | 7936222 | ||
| BP496275.1 | - | 1 | 323 | |||

Prph
Genomic Map