siRNA Details
Gene Symbol: Sdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012649.2 | NP_036781.1 | 4 | 434 | ||
GenBank mRNA | M81786.1 | - | 4 | 434 | ||
S61868.1 | - | 4 | 434 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012649.2 | NP_036781.1 | 4 | 434 | ||
GenBank mRNA | M81786.1 | - | 4 | 434 | ||
S61868.1 | - | 4 | 434 |