siRNA Details
| Gene Symbol: Sts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012661.1 | NP_036793.1 | 7 | 1525 | ||
| XM_006256960.2 | XP_006257022.1 | 7 | 1772 | |||
| XM_006256961.2 | XP_006257023.1 | 7 | 1760 | |||
| XM_006256962.2 | XP_006257024.1 | 7 | 1447 | |||
| GenBank mRNA | BC085747.1 | - | 4 | 2201 | ||
| U37138.1 | - | 7 | 1525 | |||

Sts
Genomic Map