siRNA Details
| Gene Symbol: Vamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012663.2 | NP_036795.1 | 4, 5 | 409 | ||
| GenBank mRNA | BC074003.1 | - | 4, 5 | 409 | 7097683 | |
| M24105.1 | - | 3, 4 | 347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vamp2| Gene Symbol: Vamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012663.2 | NP_036795.1 | 4, 5 | 409 | ||
| GenBank mRNA | BC074003.1 | - | 4, 5 | 409 | 7097683 | |
| M24105.1 | - | 3, 4 | 347 | |||