siRNA Details
| Gene Symbol: Pla2g16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017060.2 | NP_058756.2 | 3 | 140 | ||
| XM_006230687.2 | XP_006230749.1 | 2 | 1009 | |||
| GenBank mRNA | AB298805.1 | - | 1 | 59 | ||
| CB613004.1 | - | 3 | 140 | |||
| FQ224126.1 | - | 3 | 200 | |||

Pla2g16
Genomic Map