siRNA Details
| Gene Symbol: Psmb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D10729.1 | - | 3 | 332 | ||
| KC222892.1 | - | 3 | 346 | |||
| KC222893.1 | - | 3 | 346 | |||
| KC222894.1 | - | 3 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb8| Gene Symbol: Psmb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D10729.1 | - | 3 | 332 | ||
| KC222892.1 | - | 3 | 346 | |||
| KC222893.1 | - | 3 | 346 | |||
| KC222894.1 | - | 3 | 346 | |||