siRNA Details
| Gene Symbol: Lta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080769.2 | NP_542947.1 | 4 | 363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lta| Gene Symbol: Lta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080769.2 | NP_542947.1 | 4 | 363 | ||