siRNA Details
| Gene Symbol: Semg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012710.2 | NP_036842.2 | 1 | 67 | ||
| GenBank mRNA | BC061977.1 | - | 1 | 67 | 5623736 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Semg1| Gene Symbol: Semg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012710.2 | NP_036842.2 | 1 | 67 | ||
| GenBank mRNA | BC061977.1 | - | 1 | 67 | 5623736 | |