siRNA Details
| Gene Symbol: Chrnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052806.2 | NP_434693.1 | 5 | 511 | ||
| GenBank mRNA | AY574260.1 | - | 5 | 511 | ||
| U42976.1 | - | 571 | ||||
| X15834.1 | - | 623 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrnb4| Gene Symbol: Chrnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052806.2 | NP_434693.1 | 5 | 511 | ||
| GenBank mRNA | AY574260.1 | - | 5 | 511 | ||
| U42976.1 | - | 571 | ||||
| X15834.1 | - | 623 | ||||