siRNA Details
| Gene Symbol: Aanat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012818.2 | NP_036950.1 | 4 | 608 | ||
| XM_006247792.1 | XP_006247854.1 | 4 | 1604 | |||
| XM_006247793.1 | XP_006247855.1 | 4 | 1599 | |||
| GenBank mRNA | DQ075321.1 | - | 4 | 796 | ||
| U29664.1 | - | 3 | 397 | |||
| U38306.1 | - | 4 | 608 | |||
| U40803.1 | - | 4 | 695 | |||

Aanat
Genomic Map