siRNA Details
| Gene Symbol: Aanat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012818.2 | NP_036950.1 | 3, 4 | 423 | ||
| XM_006247792.1 | XP_006247854.1 | 3, 4 | 1419 | |||
| XM_006247793.1 | XP_006247855.1 | 3, 4 | 1414 | |||
| GenBank mRNA | DQ075321.1 | - | 3, 4 | 611 | ||
| U29664.1 | - | 2, 3 | 212 | |||
| U38306.1 | - | 3, 4 | 423 | |||
| U40803.1 | - | 3, 4 | 510 | |||

Aanat
Genomic Map