siRNA Details
| Gene Symbol: Ncl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012749.2 | NP_036881.2 | 7, 8 | 1304 | ||
| GenBank mRNA | BC085751.1 | - | 7, 8 | 1304 | 7109097 | |
| FJ817497.1 | - | 7, 8 | 743 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncl| Gene Symbol: Ncl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012749.2 | NP_036881.2 | 7, 8 | 1304 | ||
| GenBank mRNA | BC085751.1 | - | 7, 8 | 1304 | 7109097 | |
| FJ817497.1 | - | 7, 8 | 743 | |||