siRNA Details
| Gene Symbol: Inhbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080771.1 | NP_542949.1 | 2 | 492 | ||
| GenBank mRNA | AF478684.1 | - | 1 | 23 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inhbb| Gene Symbol: Inhbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080771.1 | NP_542949.1 | 2 | 492 | ||
| GenBank mRNA | AF478684.1 | - | 1 | 23 | ||