siRNA Details
| Gene Symbol: Aqp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012779.1 | NP_036911.1 | 4 | 771 | ||
| GenBank mRNA | BC078904.1 | - | 4 | 765 | ||
| BC092572.1 | - | 4 | 760 | 7122974 | ||
| BC110045.1 | - | 4 | 776 | 7100092 | ||
| U16245.1 | - | 4 | 771 | |||

Aqp5
Genomic Map