siRNA Details
Gene Symbol: Tcf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032397.1 | NP_001027569.1 | 2 | 716 | ||
GenBank mRNA | BC100106.1 | - | 2 | 716 | 7379660 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tcf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032397.1 | NP_001027569.1 | 2 | 716 | ||
GenBank mRNA | BC100106.1 | - | 2 | 716 | 7379660 |