siRNA Details
| Gene Symbol: Cebpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012831.1 | NP_036963.1 | 2 | 429 | ||
| XM_008759112.1 | XP_008757334.1 | 3 | 672 | |||
| GenBank mRNA | BC166589.1 | - | 2 | 418 | 7934938 | |
| X64403.1 | - | 2 | 429 | |||

Cebpg
Genomic Map