siRNA Details
| Gene Symbol: Sod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012880.1 | NP_037012.1 | 2 | 242 | ||
| XM_006251030.2 | XP_006251092.1 | 2 | 288 | |||
| GenBank mRNA | BC061861.1 | - | 2 | 193 | 5598594 | |
| X68041.1 | - | 2 | 217 | |||
| X94371.1 | - | 2 | 210 | |||
| Z24721.1 | - | 2 | 242 | |||

Sod3
Genomic Map