siRNA Details
| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012703.2 | NP_036835.2 | 1 | 423 | ||
| GenBank mRNA | BC089061.1 | - | 1 | 423 | 7308757 | |
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Thrsp
Thrsp| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012703.2 | NP_036835.2 | 1 | 423 | ||
| GenBank mRNA | BC089061.1 | - | 1 | 423 | 7308757 | |
 Back To Top
 Back To Top