siRNA Details
| Gene Symbol: Adora2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053294.4 | NP_445746.3 | 2 | 518 | ||
| GenBank mRNA | AF115508.1 | - | 1 | 24 | ||
| AF228684.1 | - | 1 | 124 | |||
| BC081727.1 | - | 2 | 518 | 7121208 | ||
| L08102.1 | - | 1 | 90 | |||
| M91214.2 | - | 1 | 375 | |||
| S47609.1 | - | 1 | 164 | |||
More Information
Set Membership: |
|

Adora2a
Genomic Map