siRNA Details
| Gene Symbol: Prkag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013010.2 | NP_037142.1 | 7 | 396 | ||
| XM_006257331.2 | XP_006257393.1 | 7 | 596 | |||
| GenBank mRNA | BC085749.1 | - | 6 | 382 | ||
| BC097940.1 | - | 7 | 396 | 7454478 | ||
| U42413.1 | - | 6 | 359 | |||
| X95578.1 | - | 7 | 416 | |||

Prkag1
Genomic Map