siRNA Details
| Gene Symbol: Prkag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013010.2 | NP_037142.1 | 7, 8 | 429 | ||
| XM_006257331.2 | XP_006257393.1 | 7, 8 | 629 | |||
| GenBank mRNA | BC085749.1 | - | 6, 7 | 415 | ||
| BC097940.1 | - | 7, 8 | 429 | 7454478 | ||
| U42413.1 | - | 6, 7 | 392 | |||
| X95578.1 | - | 7, 8 | 449 | |||

Prkag1
Genomic Map