siRNA Details
| Gene Symbol: Sbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013024.2 | NP_037156.2 | 4 | 635 | ||
| XM_008767547.1 | XP_008765769.1 | 3 | 621 | |||
| XM_008767548.1 | XP_008765770.1 | 3 | 441 | |||
| GenBank mRNA | CA339008.1 | - | 4 | 635 | ||
| J02675.1 | - | 3 | 645 | |||

Sbp
Genomic Map